Order Entry
Poland
ContactUsLinkComponent
Anti-FGF21 Rabbit Polyclonal Antibody
Anti-FGF21 Rabbit Polyclonal Antibody
  BSBTPA1673
undefined
Anti-FGF21 Rabbit Polyclonal Antibody
  BSBTPA1673

 

  • Antibody type:
    Primary
  • Antigen name:
    Fibroblast growth factor 21
  • Antigen symbol:
    FGF21
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    43 to 58
  • UniProtKB:
    Q9NSA1
  • Sequence:
    QVRQRYLYTDDAQQTE
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human FGF21 (43-58aa QVRQRYLYTDDAQQTE), different from the related rat and mouse sequences by two amino acids.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Fibroblast growth factor 21(FGF21) detection. Tested with WB in Human;Mouse;Rat.

FGF21(Fibroblast growth factor 21) is a protein that in humans is encoded by the FGF21 gene, which is also a member of the fibroblast growth factor(FGF) family. The FGF21 gene is mapped on 19q13.33. Using RT-PCR, Fgf21 was expressed in several types of adipose tissue in mice, including subcutaneous and epididymal fat pads and brown adipose tissue. The level of Fgf21 expression in adipose tissue was comparable to that in liver. FGF21 stimulates glucose uptake in adipocytes but not in other cell types. This effect is additive to the activity of insulin. FGF21 treatment of adipocytes is associated with phosphorylation of FRS2, a protein linking FGF receptors to the Ras/MAP kinase pathway. FGF21 also protects animals from diet-induced obesity when overexpressed in transgenic mice and lowers blood glucose and triglyceride levels when administered to diabetic rodents. Changes in Fgf21 expression due to suckling or nutritional manipulations were associated with changes in circulating free fatty acid and ketone body levels. In differentiated mouse brown adipocytes in culture, Fgf21 treatment increased the expression of thermogenic genes, caused higher total and uncoupled respiration, and enhanced glucose oxidation.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: FGF21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human