Order Entry
Poland
ContactUsLinkComponent
Anti-Monoamine Oxidase B Rabbit Polyclonal Antibody
Anti-Monoamine Oxidase B Rabbit Polyclonal Antibody
Catalog # BSBTPA1649
Supplier:  Boster Bio
undefined
Anti-Monoamine Oxidase B Rabbit Polyclonal Antibody
Catalog # BSBTPA1649
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Monoamine Oxidase B
  • Antigen symbol:
    MAOB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    HGNC:6834|monoamine oxidase B |MAOB|platelet|RP1-201D17__B.1|MAO|brain|adrenalin oxidase|amine oxidase (flavin-containing)|MGC26382|tyramine oxidase
  • Amino acid number:
    42 - 56
  • Sequence:
    RTYTIRNKNVKYVDL
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of mouse Monoamine Oxidase B (42-56aa RTYTIRNKNVKYVDL), identical to the related rat sequence.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Amine oxidase[flavin-containing] B(MAOB) detection. Tested with WB, IHC-P in Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Monoamine Oxidase B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat