Order Entry
Poland
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (CF™ 543)
Anti-IgG Goat Polyclonal Antibody (CF™ 543)
Supplier:  Biotium
Certificates
Anti-IgG Goat Polyclonal Antibody (CF™ 543)
Supplier:  Biotium

Specifications

  • Catalog No:
  • 20299-1
  • 20299.
  • Antigen symbol:
  • IgG
  • IgG
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • CF® 543
  • CF® 543
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Goat
  • Goat
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together