Order Entry
Poland
ContactUsLinkComponent
Anti-KMT5A / SETD8 / Pr-SET7 Rabbit Polyclonal Antibody
Anti-KMT5A / SETD8 / Pr-SET7 Rabbit Polyclonal Antibody
Catalog # ABCAAB111691-50
Supplier:  Abcam
undefined
Anti-KMT5A / SETD8 / Pr-SET7 Rabbit Polyclonal Antibody
Catalog # ABCAAB111691-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SET domain containing (lysine methyltransferase) 8
  • Antigen symbol:
    SETD8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    1202
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Rabbit polyclonal to KMT5A / SETD8 / Pr-SET7.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SETD8 (SET domain containing (lysine methyltransferase) 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human