Anti-Integrin beta 3 Mouse Monoclonal Antibody [clone: VI-PL2]
ABCAAB110131-50
: Abcam
- Antibody type:Primary
- Antigen name:integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61)
- Antigen symbol:ITGB3
- Clonality:Monoclonal
- Clone:VI-PL2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Seq:1202
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pack type:Vial
- Pk:50 µG
Type: Primary
Antigen: ITGB3 (integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61))
Clonality: monoclonal
Clone: VI-PL2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human