Order Entry
Poland
ContactUsLinkComponent
Anti-MMP2 Mouse Monoclonal Antibody [clone: 6E3F8]
Anti-MMP2 Mouse Monoclonal Antibody [clone: 6E3F8]
Catalog # ABCAAB86607-100
Supplier:  Abcam
undefined
Anti-MMP2 Mouse Monoclonal Antibody [clone: 6E3F8]
Catalog # ABCAAB86607-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase)
  • Antigen symbol:
    MMP2
  • Clonality:
    Monoclonal
  • Clone:
    6E3F8
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [6E3F8] to MMP2.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MMP2
Clonality: Monoclonal
Clone: 6E3F8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat