Order Entry
Poland
ContactUsLinkComponent
Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [clone: MEM-136] (PE (Phycoerythrin))
Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [clone: MEM-136] (PE (Phycoerythrin))
Catalog # ABCAAB77134-100
Dostawca:  Abcam
undefined
Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [clone: MEM-136] (PE (Phycoerythrin))
Catalog # ABCAAB77134-100
Dostawca:  Abcam

Dane techniczne

  • Antibody type:
    Primary
  • Antigen symbol:
    HLA DP DR
  • Clonality:
    Monoclonal
  • Clone:
    MEM-136
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Seq:
    0790
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 Tests
  • Pack type:
    Vial
  • Pk:
    100 Tests

Dane techniczne

Informacje o tym produkcie

PE Mouse monoclonal [MEM-136] to HLA DR + HLA DP.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: HLA DR + HLA DP
Clonality: Monoclonal
Clone: MEM-136
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human