Order Entry
Poland
ContactUsLinkComponent
Anti-CAMKIV (phospho T196 + T200) Rabbit Polyclonal Antibody
Anti-CAMKIV (phospho T196 + T200) Rabbit Polyclonal Antibody
Catalog # ABCAAB59424-100
Supplier:  Abcam
undefined
Anti-CAMKIV (phospho T196 + T200) Rabbit Polyclonal Antibody
Catalog # ABCAAB59424-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    calcium/calmodulin-dependent Protein kinase IV
  • Antigen symbol:
    CAMK4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal to CAMKIV (phospho T196 + T200).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CAMKIV
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human