Order Entry
Poland
ContactUsLinkComponent
Anti-A2B5 Mouse Monoclonal Antibody [clone: 105]
Anti-A2B5 Mouse Monoclonal Antibody [clone: 105]
  ABCAAB53521-100
 :  Abcam
undefined
Anti-A2B5 Mouse Monoclonal Antibody [clone: 105]
  ABCAAB53521-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit beta isoform
  • Antigen symbol:
    A2B5
  • Clonality:
    Monoclonal
  • Clone:
    105
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Chicken,
    Human,
    Rat,
    Mouse
  • Seq:
    1286
  • Form:
    Liquid
  • Antigen synonyms:
    PP2A B subunit isoform B'-beta|PP2A B subunit isoform B56-beta|PP2A B subunit isoform PR61-beta|PP2A B subunit isoform R5-beta|PPP2R5B
  • Molecular weight:
    57393 Da
  • Storage temperature:
    Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C or –80 °C. Avoid freeze / thaw cycle.
  • Shipping temperature:
    4 °C
  • Immunogen:
    The exact immunogen used to generate this antibody is proprietary information.
  • Tested applications:
    Flow Cyt (Intra), ICC/IF
  • Purification:
    IgM fraction
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [105] to A2B5.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: A2B5
Clonality: Monoclonal
Clone: 105
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Mouse; Rat; Human