Order Entry
Poland
ContactUsLinkComponent
Anti-STAT5 (phospho Y694) Rabbit Monoclonal Antibody [clone: E208]
Anti-STAT5 (phospho Y694) Rabbit Monoclonal Antibody [clone: E208]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-STAT5 (phospho Y694) Rabbit Monoclonal Antibody [clone: E208]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB32364-100
  • ABCAAB32364-1ML
  • ABCAAB32364-20UL
  • Antigen symbol:
  • STAT5A
  • Antigen name:
  • Signal transducer and activator of transcription 5A
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • E208
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together