Order Entry
Poland
ContactUsLinkComponent
Anti-C Reactive Protein Mouse Monoclonal Antibody [clone: C6] (HRP (Horseradish Peroxidase))
Anti-C Reactive Protein Mouse Monoclonal Antibody [clone: C6] (HRP (Horseradish Peroxidase))
Catalog # ABCAAB24462-200
Supplier:  Abcam
undefined
Anti-C Reactive Protein Mouse Monoclonal Antibody [clone: C6] (HRP (Horseradish Peroxidase))
Catalog # ABCAAB24462-200
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C Reactive Protein
  • Antigen symbol:
    CRP
  • Clonality:
    Monoclonal
  • Clone:
    C6
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µg
  • Pack type:
    Vial
  • Pk:
    200 µG

Specifications

About this item

HRP Mouse monoclonal [C6] to C Reactive Protein.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: C Reactive Protein
Clonality: monoclonal
Clone: C6
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human