Order Entry
Poland
ContactUsLinkComponent
Anti-CD2 Mouse Monoclonal Antibody [clone: MEM-65] (Biotinylated)
Anti-CD2 Mouse Monoclonal Antibody [clone: MEM-65] (Biotinylated)
Catalog # ABCAAB21901-100
Supplier:  Abcam
undefined
Anti-CD2 Mouse Monoclonal Antibody [clone: MEM-65] (Biotinylated)
Catalog # ABCAAB21901-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD2 molecule
  • Antigen symbol:
    CD2
  • Clonality:
    Monoclonal
  • Clone:
    MEM-65
  • Conjugation:
    Biotin
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Biotin Mouse monoclonal [MEM-65] to CD2.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: MEM-65
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human