Anti-HLA Class II DRB1 Mouse Monoclonal Antibody [clone: DA2]
Catalog # ABCAAB20184-50
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen symbol:HLA DR beta
- Clonality:Monoclonal
- Clone:DA2
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Seq:1202
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pack type:Vial
- Pk:50 µG
Specifications
About this item
Mouse monoclonal [DA2] to HLA Class II DRB1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: HLA Class II DRB1
Clonality: Monoclonal
Clone: DA2
Conjugation:
Epitope: 1st domain, monomorphic.
Host: Mouse
Isotype: IgG1
Reactivity: Human