Order Entry
Poland
ContactUsLinkComponent
 
Anti-Prion protein PrP Mouse Monoclonal Antibody [clone: 7B6/D2]
Catalog # ABCAAB2882-100
Supplier:  Abcam
undefined
Anti-Prion protein PrP Mouse Monoclonal Antibody [clone: 7B6/D2]
Catalog # ABCAAB2882-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Prion Protein PrP
  • Clonality:
    Monoclonal
  • Clone:
    7B6+D2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Bovine
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [7B6/D2] to Prion protein PrP.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Prion protein PrP
Clonality: Monoclonal
Clone: 7B6 / D2
Conjugation:
Epitope: This antibody has been shown to specifically bind between amino acids 123 - 136 and 140 - 160 of bovine prion protein, determined by pepscan (Osman, A.A. et al.).
Host: Mouse
Isotype: IgG1
Reactivity: Bovine