Anti-HLA-DR Mouse Monoclonal Antibody [clone: LN3] (FITC (Fluorescein Isothiocyanate))
ABCAAB1182-500
: Abcam
- Antibody type:Primary
- Antigen symbol:HLA DR
- Clonality:Monoclonal
- Clone:LN3
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Seq:1418
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µl
- Pack type:Vial
- Pk:500 µl
FITC Mouse monoclonal [LN3] to HLA-DR.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: LN3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human