Anti-TUBB2B Rabbit Polyclonal Antibody
ABNOH00347733-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Tubulin beta 2B
- Antigen symbol:TUBB2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:347733
- Antigen synonyms:bA506K6.1|PMGYSA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TUBB2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human