Anti-IFNE Mouse Monoclonal Antibody [clone: 2F8]
ABNOH00338376-M02
: Abnova
- Antibody type:Primary
- Antigen name:Interferon epsilon
- Antigen symbol:IFNE
- Clonality:Monoclonal
- Clone:2F8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:338376
- Antigen synonyms:IFN-E|PRO655|IFNT1|INFE1|IFNE1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: IFNE
Clonality: Monoclonal
Clone: 2F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human