Anti-DCST1 Mouse Polyclonal Antibody
50 µl
Catalog # ABNOH00149095-A01
Supplier: Abnova
Jump to:
Product Details & Documents
Type: Primary
Antigen: DCST1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Documents
TDS_ABNOH00149095-A01.pdf
Datasheet
Specifications
- Size:50 µl
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Storage temperature:Cool
- Host:Mouse
- Cross adsorption:No
- Shipping temperature:Ice
- Antigen name:DC-STAMP Domain Containing 1
- Cat. No.:ABNOH00149095-A01
- Reactivity:Human
- Gene ID:149095
- Conjugation:Unconjugated
- Antigen symbol:DCST1
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



