Anti-GSC Mouse Monoclonal Antibody [clone: 3C7]
Catalog # ABNOH00145258-M11
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Goosecoid homeobox
- Antigen symbol:GSC
- Clonality:Monoclonal
- Clone:3C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:145258
- Antigen synonyms:SAMS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GSC
Clonality: Monoclonal
Clone: 3C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human