Order Entry
Poland
ContactUsLinkComponent
Anti-GSC Mouse Monoclonal Antibody [clone: 4B9]
Anti-GSC Mouse Monoclonal Antibody [clone: 4B9]
Catalog # ABNOH00145258-M10
Supplier:  Abnova
undefined
Anti-GSC Mouse Monoclonal Antibody [clone: 4B9]
Catalog # ABNOH00145258-M10
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Goosecoid homeobox
  • Antigen symbol:
    GSC
  • Clonality:
    Monoclonal
  • Clone:
    4B9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    145258
  • Antigen synonyms:
    SAMS
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: GSC
Clonality: Monoclonal
Clone: 4B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human