Anti-CALR3 Mouse Monoclonal Antibody [clone: 4E3]
ABNOH00125972-M01
: Abnova
- Antibody type:Primary
- Antigen name:Calreticulin 3
- Antigen symbol:CALR3
- Clonality:Monoclonal
- Clone:4E3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:125972
- Antigen synonyms:CRT2|CMH19|CT93
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CALR3
Clonality: Monoclonal
Clone: 4E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human