Anti-NXNL1 Mouse Monoclonal Antibody [clone: 7H3]
Catalog # ABNOH00115861-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Nucleoredoxin-like 1
- Antigen symbol:NXNL1
- Clonality:Monoclonal
- Clone:7H3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:115861
- Antigen synonyms:TXNL6|RDCVF
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: NXNL1
Clonality: Monoclonal
Clone: 7H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human