Anti-TNFRSF13C Rabbit Polyclonal Antibody
ABNOH00115650-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Receptor Superfamily Member 13C
- Antigen symbol:TNFRSF13C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:115650
- Antigen synonyms:CVID4|BAFFR|BAFF-R|BROMIX|CD268|Signal Transducing Adaptor Molecule (SH3 Domain And ITAM Motif) 1 And 2|prolixin
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TNFRSF13C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse