Anti-CAPZA3 Rabbit Polyclonal Antibody
Catalog # ABNOH00093661-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Capping protein (actin filament) muscle Z-line, alpha 3
- Antigen symbol:CAPZA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:93661
- Antigen synonyms:HEL-S-86|Gsg3|CAPPA3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CAPZA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human