Anti-COL23A1 Mouse Monoclonal Antibody [clone: 2C9]
Catalog # ABNOH00091522-M01A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Collagen, type XXIII, alpha 1
- Antigen symbol:COL23A1
- Clonality:Monoclonal
- Clone:2C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:91522
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: COL23A1
Clonality: Monoclonal
Clone: 2C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human