Anti-APOL3 Mouse Polyclonal Antibody
ABNOH00080833-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Apolipoprotein L3
- Antigen symbol:APOL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:80833
- Antigen synonyms:CG12_1|CG121|apoL-III|APOLIII
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: APOL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human