Anti-CERS4 Mouse Monoclonal Antibody [clone: 7D5]
ABNOH00079603-M03
: Abnova
- Antibody type:Primary
- Antigen name:Ceramide synthase 4
- Antigen symbol:CERS4
- Clonality:Monoclonal
- Clone:7D5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:79603
- Antigen synonyms:Trh1|LASS4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CERS4
Clonality: Monoclonal
Clone: 7D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human