Anti-SECISBP2 Mouse Polyclonal Antibody
Catalog # ABNOH00079048-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:SECIS binding protein 2
- Antigen symbol:SECISBP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:79048
- Antigen synonyms:SBP2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: SECISBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human