Anti-POGK Rabbit Polyclonal Antibody
ABNOH00057645-D01
: Abnova
- Antibody type:Primary
- Antigen name:Pogo Transposable Element With KRAB Domain
- Antigen symbol:POGK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:57645
- Antigen synonyms:BASS2|LST003|KRBOX2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: POGK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human