Anti-SPHK2 Rabbit Polyclonal Antibody
Catalog # ABNOH00056848-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Sphingosine Kinase 2
- Antigen symbol:SPHK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:56848
- Antigen synonyms:SK-2|SK 2|SPHK2|Sphingosine kinase 2|SPK-2|SPK 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SPHK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human