Anti-TUBB7P Rabbit Polyclonal Antibody
Catalog # ABNOH00056604-D01
Dostawca: Abnova
Dane techniczne
- Antibody type:Primary
- Antigen name:Tubulin beta 7
- Antigen symbol:TUBB7P
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:56604
- Antigen synonyms:TUBB4Q
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Dane techniczne
Informacje o tym produkcie
Type: Primary
Antigen: TUBB7P
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human