Anti-DMAP1 Mouse Polyclonal Antibody
Catalog # ABNOH00055929-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:DNA methyltransferase 1 associated protein 1
- Antigen symbol:DMAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:55929
- Antigen synonyms:DNMAP1|FLJ11543|DKFZp686L09142|SWC4|MEAF2|DNMTAP1|KIAA1425|EAF2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: DMAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human