Anti-TRIM36 Mouse Monoclonal Antibody [clone: 2D11]
Catalog # ABNOH00055521-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tripartite Motif Containing 36
- Antigen symbol:TRIM36
- Clonality:Monoclonal
- Clone:2D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:55521
- Antigen synonyms:HAPRIN|RBCC728|RNF98
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TRIM36
Clonality: Monoclonal
Clone: 2D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human