Anti-CSGALNACT2 Rabbit Polyclonal Antibody
Catalog # ABNOH00055454-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Chondroitin sulfate N-acetylgalactosaminyltransferase 2
- Antigen symbol:CSGALNACT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:55454
- Antigen synonyms:GALNACT-2|CHGN2|GALNACT2|PRO0082
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CSGALNACT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human