Anti-RCBTB1 Mouse Monoclonal Antibody [clone: 1E4]
ABNOH00055213-M07
: Abnova
- Antibody type:Primary
- Antigen name:RCC1 And BTB Domain Containing Protein 1
- Antigen symbol:RCBTB1
- Clonality:Monoclonal
- Clone:1E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:55213
- Antigen synonyms:CLLD7|CLLL7|GLP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: RCBTB1
Clonality: Monoclonal
Clone: 1E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human