Anti-PHIP Mouse Polyclonal Antibody
ABNOH00055023-A01
: Abnova
- Antibody type:Primary
- Antigen name:Pleckstrin Homology Domain Interacting Protein
- Antigen symbol:PHIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:55023
- Antigen synonyms:DCAF14|BRWD2|ndrp|WDR11
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: PHIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human