Anti-WDR74 Rabbit Polyclonal Antibody
ABNOH00054663-D01P
: Abnova
- Antibody type:Primary
- Antigen name:WD Repeat Domain 74
- Antigen symbol:WDR74
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:54663
- Antigen synonyms:NSA1|NOP seven-associated protein 1|WD repeat-containing protein 74
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: WDR74
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human