Anti-DDX56 Mouse Polyclonal Antibody
Catalog # ABNOH00054606-B01P
Dostawca: Abnova
Dane techniczne
- Antibody type:Primary
- Antigen name:DEAD (Asp-Glu-Ala-Asp) box helicase 56
- Antigen symbol:DDX56
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:54606
- Antigen synonyms:NOH61|DDX26|DDX21
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Dane techniczne
Informacje o tym produkcie
Type: Primary
Antigen: DDX56
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human