Order Entry
Poland
ContactUsLinkComponent
Anti-SIRT6 Mouse Monoclonal Antibody [clone: 1D8]
Anti-SIRT6 Mouse Monoclonal Antibody [clone: 1D8]
Catalog # ABNOH00051548-M01
Supplier:  Abnova
undefined
Anti-SIRT6 Mouse Monoclonal Antibody [clone: 1D8]
Catalog # ABNOH00051548-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sirtuin 6
  • Antigen symbol:
    SIRT6
  • Clonality:
    Monoclonal
  • Clone:
    1D8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Gene ID:
    51548
  • Antigen synonyms:
    SIR2-like protein 6|Mono-ADP-ribosyltransferase sirtuin-6|Silent information regulator 2-like protein 6.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: SIRT6
Clonality: Monoclonal
Clone: 1D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human