Anti-RNF181 Rabbit Polyclonal Antibody
Catalog # ABNOH00051255-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ring Finger Protein 181
- Antigen symbol:RNF181
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:51255
- Antigen synonyms:HSPC238
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: RNF181
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse