Anti-YBX2 Rabbit Polyclonal Antibody
ABNOH00051087-D01
: Abnova
- Antibody type:Primary
- Antigen name:Y Box Binding Protein 2
- Antigen symbol:YBX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:51087
- Antigen synonyms:DBPC|Y-box-binding protein 2|Dbpc|Contrin|YBX2|MSY2|CONTRIN|CSDA3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: YBX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human