Order Entry
Poland
ContactUsLinkComponent
Anti-AK3 Rabbit Polyclonal Antibody
Anti-AK3 Rabbit Polyclonal Antibody
Catalog # ABNOH00050808-D01
Supplier:  Abnova
undefined
Anti-AK3 Rabbit Polyclonal Antibody
Catalog # ABNOH00050808-D01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Adenylate Kinase 3
  • Antigen symbol:
    AK3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    50808
  • Antigen synonyms:
    AK3|mitochondrial|AK4|AK3L1|ATP-AMP transphosphorylase|Adenylate kinase 3-like|Adenylate kinase isoenzyme 4
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: AK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human