Anti-NEUROG3 Mouse Monoclonal Antibody [clone: 4B2]
Catalog # ABNOH00050674-M06
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Neurogenin 3
- Antigen symbol:NEUROG3
- Clonality:Monoclonal
- Clone:4B2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:50674
- Antigen synonyms:NGN|bHLHa7|Atoh5|NGN1|Neurogenic basic-helix-loop-helix- protein|NGN-3|ngn3|Neurogenic differentiation factor 3|NEUROG1|NeuroD3|NEUROD3|Math4B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: NEUROG3
Clonality: Monoclonal
Clone: 4B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human