Anti-CERS2 Mouse Monoclonal Antibody [clone: 4D8]
Catalog # ABNOH00029956-M10
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ceramide synthase 2
- Antigen symbol:CERS2
- Clonality:Monoclonal
- Clone:4D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:29956
- Antigen synonyms:SP260|L3|TMSG1|LASS2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CERS2
Clonality: Monoclonal
Clone: 4D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human