Anti-CERS2 Mouse Monoclonal Antibody [clone: 1A6]
ABNOH00029956-M01A
: Abnova
- Antibody type:Primary
- Antigen name:Ceramide synthase 2
- Antigen symbol:CERS2
- Clonality:Monoclonal
- Clone:1A6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:29956
- Antigen synonyms:SP260|L3|TMSG1|LASS2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: CERS2
Clonality: Monoclonal
Clone: 1A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human