Anti-ATAD2 Mouse Polyclonal Antibody
ABNOH00029028-B01P
: Abnova
- Antibody type:Primary
- Antigen name:ATPase Family, AAA Domain Containing 2
- Antigen symbol:ATAD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:29028
- Antigen synonyms:PRO2000|ANCCA|CT137
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: ATAD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human