Anti-HTRA2 Mouse Monoclonal Antibody [clone: 4F10]
Catalog # ABNOH00027429-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:High Temperature Requirement Protein A2
- Antigen symbol:HTRA2
- Clonality:Monoclonal
- Clone:4F10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:27429
- Antigen synonyms:mitochondrial|Serine protease HTRA2|OMI|HtrA2|PRSS25|PARK13|High temperature requirement protein A2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: HTRA2
Clonality: Monoclonal
Clone: 4F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human