Anti-FBXO10 Mouse Polyclonal Antibody
ABNOH00026267-A01
: Abnova
- Antibody type:Primary
- Antigen name:F-box protein 10
- Antigen symbol:FBXO10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:26267
- Antigen synonyms:PRMT11|FBX10
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: FBXO10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human