Anti-OSBPL3 Mouse Polyclonal Antibody
ABNOH00026031-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Oxysterol Binding Protein-like 3
- Antigen symbol:OSBPL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:26031
- Antigen synonyms:OSBP3|ORP3|ORP-3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: OSBPL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human