Anti-CHRDL2 Mouse Polyclonal Antibody
50 µg
Catalog # ABNOH00025884-B01P
Supplier: Abnova
Jump to:
Product Details & Documents
Type: Primary
Antigen: CHRDL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Documents
TDS_ABNOH00025884-B01P.pdf
Datasheet
Specifications
- Size:50 µg
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Storage temperature:Cool
- Host:Mouse
- Cross adsorption:No
- Shipping temperature:Ice
- Antigen name:Chordin-like 2
- Cat. No.:ABNOH00025884-B01P
- Reactivity:Human
- Gene ID:25884
- Antigen synonyms:CHL2|Chordin related protein 2|Breast tumor novel factor 1|CRDL2_HUMAN.|Chordin like protein 2 precursor|BNF1|chordin like 2
- Conjugation:Unconjugated
- Antigen symbol:CHRDL2



